siRNA Details
| Gene Symbol: SCNN1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131558.1 | - | 1 | 85 | ||
| BX424308.2 | - | 2 | 144 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453732.1 | - | 2 | 142 | Chr. 1 - 1275237 - 1279393 [+] on Build GRCh38 | |

SCNN1D
Genomic Map