siRNA Details
| Gene Symbol: SERPINB9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033851.1 | 2 | 882 | |||
| GenBank mRNA | BC025340.1 | - | 2 | 882 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420981.2 | - | 2 | 882 | Chr. 6 - 2854926 - 2876510 [-] on Build GRCh38 | |

SERPINB9P1
Genomic Map