siRNA Details
| Gene Symbol: MKX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121652.1 | 2 | 505 | |||
| GenBank mRNA | BG194628.1 | - | 1 | 177 | ||
| BU195543.1 | - | 2 | 342 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419777.1 | - | 2 | 334 | Chr. 10 - 27744786 - 27767794 [+] on Build GRCh38 | |

MKX-AS1
Genomic Map