siRNA Details
| Gene Symbol: P2RX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029075.1 | XP_016884564.1 | 1 | 69 | ||
| Gene Symbol: THAP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027051.1 | 2 | 2009 | |||
| GenBank mRNA | AK096901.1 | - | 2 | 1714 | ||
| AK097739.1 | - | 2 | 1277 | |||

P2RX6
Genomic Map