siRNA Details
| Gene Symbol: LOC101929441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_123739.1 | 2 | 699 | |||
| GenBank mRNA | BC038769.1 | - | 2 | 699 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433869.1 | - | 2 | 699 | Chr. 1 - 204141408 - 204143009 [+] on Build GRCh38 | |

LOC101929441
Genomic Map