siRNA Details
Gene Symbol: LOC101929441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123739.1 | 2 | 261 | |||
GenBank mRNA | BC038769.1 | - | 2 | 261 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000433869.1 | - | 2 | 261 | Chr. 1 - 204141408 - 204143009 [+] on Build GRCh38 |
