siRNA Details
| Gene Symbol: KLF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC031250.1 | - | 7 | 2414 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000434244.5 | - | 7 | 2416 | Chr. 1 - 44043928 - 44104975 [+] on Build GRCh38 | |

KLF17
Genomic Map