siRNA Details
| Gene Symbol: LOC284600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056486.1 | - | 2 | 1000 | ||
| BC000227.1 | - | 1 | 462 | |||
| BC006442.1 | - | 1 | 462 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448179.1 | - | 2 | 1000 | Chr. 1 - 911435 - 914948 [+] on Build GRCh38 | |

LOC284600
Genomic Map