siRNA Details
Gene Symbol: LOC105376480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134497.1 | 2 | 426 | |||
GenBank mRNA | BX114033.1 | - | 2 | 426 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416310.1 | - | 2 | 394 | Chr. 10 - 30723251 - 30723740 [+] on Build GRCh38 |
