siRNA Details
| Gene Symbol: ZNF341-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110623.1 | 3 | 196 | |||
| GenBank mRNA | AA453965.1 | - | 3 | 196 | ||
| BX102976.1 | - | 3 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF341-AS1| Gene Symbol: ZNF341-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110623.1 | 3 | 196 | |||
| GenBank mRNA | AA453965.1 | - | 3 | 196 | ||
| BX102976.1 | - | 3 | 194 | |||