siRNA Details
Gene Symbol: LOC340512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126029.1 | 1 | 95 | |||
GenBank mRNA | AK097706.1 | - | 1 | 95 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000439901.1 | - | 1 | 95 | Chr. 9 - 106974833 - 107102988 [-] on Build GRCh38 |
