siRNA Details
| Gene Symbol: LINC01614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132383.1 | 2 | 414 | |||
| GenBank mRNA | AI752239.1 | - | 2 | 356 | ||
| CB044991.1 | - | 2 | 424 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415479.1 | - | 2 | 414 | Chr. 2 - 215718043 - 215719424 [+] on Build GRCh38 | |

LINC01614
Genomic Map