siRNA Details
Gene Symbol: LINC00692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034055.1 | 8 | 723 | |||
GenBank mRNA | BC033370.1 | - | 8 | 726 | ||
BC126324.1 | - | 7 | 560 | |||
BC143987.1 | - | 7 | 637 | |||
BC143998.1 | - | 7 | 634 | |||
BC143999.1 | - | 7 | 664 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000451284.5 | - | 8 | 726 | Chr. 3 - 25858533 - 25873698 [-] on Build GRCh38 |
