siRNA Details
| Gene Symbol: IPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270839.4 | XP_005270896.1 | 9 | 5052 | ||
| XM_006710623.3 | XP_006710686.1 | 8 | 4943 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000430643.1 | - | 1 | 252 | Chr. 1 - 45694684 - 45697075 [-] on Build GRCh38 | |

IPP
Genomic Map