siRNA Details
Gene Symbol: IPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270839.4 | XP_005270896.1 | 9 | 5030 | ||
XM_006710623.3 | XP_006710686.1 | 8 | 4921 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000430643.1 | - | 1 | 230 | Chr. 1 - 45694684 - 45697075 [-] on Build GRCh38 |
