siRNA Details
| Gene Symbol: LOC101928307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125872.1 | 2 | 341 | |||
| GenBank mRNA | AB073651.1 | - | 1 | 2011 | ||
| AL833240.1 | - | 1 | 2174 | |||
| BM686538.1 | - | 2 | 349 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419979.1 | - | 2 | 341 | Chr. 6 - 68632663 - 68634324 [-] on Build GRCh38 | |

LOC101928307
Genomic Map