siRNA Details
Gene Symbol: LINC01520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120671.1 | 2 | 813 | |||
GenBank mRNA | AL117638.1 | - | 2 | 823 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441457.1 | - | 2 | 813 | Chr. 10 - 85449592 - 85491996 [+] on Build GRCh38 |
