siRNA Details
| Gene Symbol: LOC101926966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110201.1 | 6 | 860 | |||
| GenBank mRNA | AK093525.1 | - | 6 | 860 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000492352.5 | - | 6 | 860 | Chr. 2 - 15690782 - 15718961 [+] on Build GRCh38 | |

LOC101926966
Genomic Map