siRNA Details
Gene Symbol: LINC01436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110419.1 | 1 | 432 | |||
GenBank mRNA | CF127993.1 | - | 1 | 414 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457157.2 | - | 1 | 414 | Chr. 21 - 36005338 - 36007838 [+] on Build GRCh38 |
