siRNA Details
| Gene Symbol: LINC01436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110419.1 | 2 | 1011 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000457157.2 | - | 2 | 993 | Chr. 21 - 36005338 - 36007838 [+] on Build GRCh38 | |

LINC01436
Genomic Map