siRNA Details
Gene Symbol: LOC101927377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134501.1 | 2 | 1155 | |||
GenBank mRNA | BC047609.1 | - | 2 | 1155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000413818.2 | - | 2 | 1155 | Chr. 20 - 47352561 - 47354633 [+] on Build GRCh38 |
