siRNA Details
Gene Symbol: LINC01468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120641.1 | 4 | 538 | |||
NR_120642.1 | 3 | 503 | ||||
GenBank mRNA | BC015429.1 | - | 3 | 503 | ||
BF981308.1 | - | 501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435813.6 | - | 3 | 503 | Chr. 10 - 52450874 - 52455272 [-] on Build GRCh38 |
