siRNA Details
| Gene Symbol: LINC01468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120641.1 | 3 | 250 | |||
| NR_120642.1 | 2 | 215 | ||||
| GenBank mRNA | BC015429.1 | - | 2 | 215 | ||
| BF981308.1 | - | 214 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01468| Gene Symbol: LINC01468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120641.1 | 3 | 250 | |||
| NR_120642.1 | 2 | 215 | ||||
| GenBank mRNA | BC015429.1 | - | 2 | 215 | ||
| BF981308.1 | - | 214 | ||||