siRNA Details
| Gene Symbol: LINC01468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120641.1 | 4 | 761 | |||
| NR_120642.1 | 3 | 726 | ||||
| GenBank mRNA | BC015429.1 | - | 3 | 726 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435813.6 | - | 3 | 726 | Chr. 10 - 52450874 - 52455272 [-] on Build GRCh38 | |

LINC01468
Genomic Map