siRNA Details
| Gene Symbol: KLF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026804.1 | 2 | 496 | |||
| GenBank mRNA | AF289565.1 | - | 2 | 59 | ||
| AK023259.1 | - | 2 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLF3-AS1| Gene Symbol: KLF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026804.1 | 2 | 496 | |||
| GenBank mRNA | AF289565.1 | - | 2 | 59 | ||
| AK023259.1 | - | 2 | 496 | |||