siRNA Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 3 | 547 | |||
| GenBank mRNA | CR745839.1 | - | 2 | 297 | ||
| DR422469.1 | - | 3 | 547 | |||
| LN812954.1 | - | 3 | 512 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000450063.1 | - | 1 | 181 | Chr. 7 - 116570960 - 116573744 [-] on Build GRCh38 | |

LINC01510
Genomic Map