siRNA Details
| Gene Symbol: ITPR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108075.1 | 2 | 828 | |||
| GenBank mRNA | AL832729.1 | - | 1 | 2253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412804.1 | - | 2 | 828 | Chr. 3 - 4490891 - 4493163 [-] on Build GRCh38 | |

ITPR1-AS1
Genomic Map