siRNA Details
| Gene Symbol: LOC101926964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110628.1 | 6 | 1534 | |||
| GenBank mRNA | AK097193.1 | - | 6 | 1537 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423403.1 | - | 6 | 1534 | Chr. 1 - 60659631 - 60825584 [-] on Build GRCh38 | |

LOC101926964
Genomic Map