siRNA Details
| Gene Symbol: APCDD1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034147.1 | 6 | 858 | |||
| GenBank mRNA | AK054637.1 | - | 6 | 860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

APCDD1L-AS1| Gene Symbol: APCDD1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034147.1 | 6 | 858 | |||
| GenBank mRNA | AK054637.1 | - | 6 | 860 | ||