siRNA Details
| Gene Symbol: APCDD1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034147.1 | 7 | 987 | |||
| GenBank mRNA | AK054637.1 | - | 7 | 989 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427140.5 | - | 7 | 987 | Chr. 20 - 58515379 - 58619888 [+] on Build GRCh38 | |

APCDD1L-AS1
Genomic Map