siRNA Details
| Gene Symbol: APCDD1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091704.1 | - | 3 | 551 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000447767.1 | - | 3 | 676 | Chr. 20 - 58515543 - 58526032 [+] on Build GRCh38 | |

APCDD1L-AS1
Genomic Map