siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125974.1 | 2 | 487 | |||
| NR_125975.1 | 4 | 671 | ||||
| NR_125976.1 | 4 | 671 | ||||
| GenBank mRNA | AK125759.1 | - | 4 | 357 | ||
| BC034623.1 | - | 4 | 337 | |||
| BC043601.1 | - | 2 | 458 | |||

LOC101929147
Genomic Map