siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125974.1 | 1 | 59 | |||
| NR_125975.1 | 1 | 59 | ||||
| NR_125976.1 | 1 | 59 | ||||
| NR_125977.1 | 1 | 59 | ||||
| GenBank mRNA | BC043601.1 | - | 1 | 30 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929147| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125974.1 | 1 | 59 | |||
| NR_125975.1 | 1 | 59 | ||||
| NR_125976.1 | 1 | 59 | ||||
| NR_125977.1 | 1 | 59 | ||||
| GenBank mRNA | BC043601.1 | - | 1 | 30 | ||