siRNA Details
Gene Symbol: LINC00689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024394.1 | 1 | 994 | |||
GenBank mRNA | BC042556.1 | - | 1 | 928 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024394.1 | 1 | 994 | |||
GenBank mRNA | BC042556.1 | - | 1 | 928 |