siRNA Details
| Gene Symbol: LOC100132735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038399.1 | 3 | 262 | |||
| GenBank mRNA | BC039503.1 | - | 3 | 291 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431609.1 | - | 3 | 244 | Chr. 6 - 139854919 - 139860151 [+] on Build GRCh38 | |

LOC100132735
Genomic Map