siRNA Details
| Gene Symbol: LOC643355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270945.1 | NP_001257874.1 | 2 | 1745 | ||
| GenBank mRNA | BC034233.1 | - | 2 | 1777 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000438293.1 | - | 2 | 1777 | Chr. 1 - 111990536 - 111998842 [+] on Build GRCh38 | |

LOC643355
Genomic Map