siRNA Details
| Gene Symbol: LINC01305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038897.1 | 3 | 2664 | |||
| GenBank mRNA | AL833646.1 | - | 3 | 2664 | ||
| BC043589.1 | - | 2 | 1277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01305| Gene Symbol: LINC01305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038897.1 | 3 | 2664 | |||
| GenBank mRNA | AL833646.1 | - | 3 | 2664 | ||
| BC043589.1 | - | 2 | 1277 | |||