siRNA Details
Gene Symbol: LINC01353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117097.1 | 2 | 268 | |||
NR_117098.1 | 2 | 240 | ||||
GenBank mRNA | CK902257.1 | - | 2 | 240 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117097.1 | 2 | 268 | |||
NR_117098.1 | 2 | 240 | ||||
GenBank mRNA | CK902257.1 | - | 2 | 240 |