siRNA Details
| Gene Symbol: LINC00278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046502.1 | 1 | 207 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425031.1 | - | 1 | 205 | Chr. Y - 3002998 - 3102272 [+] on Build GRCh38 | |

LINC00278
Genomic Map