siRNA Details
Gene Symbol: LOC400002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135047.1 | 3 | 3085 | |||
GenBank mRNA | AK128230.1 | - | 3 | 3086 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000420905.1 | - | 3 | 3084 | Chr. 12 - 9948137 - 9953336 [-] on Build GRCh38 |
