siRNA Details
Gene Symbol: KCCAT211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110826.1 | 3 | 1608 | |||
GenBank mRNA | AK056252.1 | - | 3 | 1609 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000436078.5 | - | 3 | 1608 | Chr. 3 - 177816865 - 177899224 [+] on Build GRCh38 |
