siRNA Details
| Gene Symbol: LOC101927596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135584.1 | 2 | 438 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449346.1 | - | 2 | 438 | Chr. 2 - 197197991 - 197199273 [+] on Build GRCh38 | |

LOC101927596
Genomic Map