siRNA Details
| Gene Symbol: DLG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046586.1 | 2 | 405 | |||
| NR_109801.1 | 2 | 372 | ||||
| GenBank mRNA | BX102208.1 | - | 2 | 405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLG3-AS1| Gene Symbol: DLG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046586.1 | 2 | 405 | |||
| NR_109801.1 | 2 | 372 | ||||
| GenBank mRNA | BX102208.1 | - | 2 | 405 | ||