siRNA Details
| Gene Symbol: LOC646268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033946.1 | 3 | 394 | |||
| GenBank mRNA | BC029599.1 | - | 3 | 393 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414848.1 | - | 3 | 393 | Chr. 1 - 158132044 - 158140639 [-] on Build GRCh38 | |

LOC646268
Genomic Map