siRNA Details
| Gene Symbol: LINC00410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027039.1 | 6 | 1329 | |||
| GenBank mRNA | AK055781.1 | - | 6 | 1328 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419272.1 | - | 6 | 1329 | Chr. 13 - 90890954 - 90926597 [-] on Build GRCh38 | |

LINC00410
Genomic Map