siRNA Details
| Gene Symbol: LOC101927136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110840.1 | 4 | 1044 | |||
| GenBank mRNA | AK125128.1 | - | 4 | 1254 | ||
| AK126334.1 | - | 5 | 1119 | |||
| BC050580.1 | - | 4 | 1020 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927136| Gene Symbol: LOC101927136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110840.1 | 4 | 1044 | |||
| GenBank mRNA | AK125128.1 | - | 4 | 1254 | ||
| AK126334.1 | - | 5 | 1119 | |||
| BC050580.1 | - | 4 | 1020 | |||