siRNA Details
| Gene Symbol: LINC00899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027036.1 | 3 | 411 | |||
| GenBank mRNA | AI760841.1 | - | 3 | 396 | ||
| AL137569.1 | - | 1 | 1438 | |||
| BC037213.1 | - | 2 | 1744 | |||
| CD674752.1 | - | 3 | 328 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421538.1 | - | 2 | 1744 | Chr. 22 - 46040266 - 46044610 [-] on Build GRCh38 | |

LINC00899
Genomic Map