siRNA Details
| Gene Symbol: ABCA9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126414.1 | 2 | 151 | |||
| GenBank mRNA | CB960713.1 | - | 178 | |||
| CB961243.1 | - | 178 | ||||
| DA128020.1 | - | 2 | 163 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000458677.1 | - | 2 | 151 | Chr. 17 - 69004714 - 69018323 [+] on Build GRCh38 | |

ABCA9-AS1
Genomic Map