siRNA Details
| Gene Symbol: CHODL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024354.1 | 3 | 407 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000447175.1 | - | 3 | 407 | Chr. 21 - 17835016 - 17885608 [-] on Build GRCh38 | |

CHODL-AS1
Genomic Map