siRNA Details
Gene Symbol: LINC01354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038426.1 | 3 | 1787 | |||
GenBank mRNA | BC040195.2 | - | 3 | 1784 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435574.1 | - | 3 | 1787 | Chr. 1 - 234527891 - 234531779 [-] on Build GRCh38 |
